missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOT Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809573
This item is not returnable.
View return policy
Description
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| MYOT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UBF9 | |
| MYOT | |
| Full length human recombit protein of human MYOT produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3D1 | |
| Unconjugated | |
| MYOT | |
| LGMD1, LGMD1A, MFM3, TTID, TTOD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9499 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction