Learn More
MYOT Mouse anti-Human, Clone: OTI10A11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809553
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| MYOT | |
| Monoclonal | |
| Unconjugated | |
| MYOT | |
| LGMD1, LGMD1A, MFM3, TTID, TTOD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9499 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI10A11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UBF9 | |
| MYOT | |
| Full length human recombit protein of human MYOT produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.