Learn More
MYOT Monoclonal Antibody (OTI8A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809549
Encompass_Preferred
 View more versions of this product
Product Code. 17501914
  • € 540.87 / 100µL
Description
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
Specifications
MYOT
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UBF9
MYOT
Full length human recombit protein of human MYOT produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI8A7
Unconjugated
MYOT
LGMD1, LGMD1A, MFM3, TTID, TTOD
Mouse
Affinity Chromatography
RUO
9499
-20° C, Avoid Freeze/Thaw Cycles