missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOZ1 Monoclonal Antibody (OTI8A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808907
This item is not returnable.
View return policy
Description
Myozenins may serve as intracellular binding proteins involved in linking Z-disk proteins such as alpha-actinin, gamma-filamin, TCAP/telethonin, LDB3/ZASP and localizing calcineurin signaling to the sarcomere. This protein plays an important role in the modulation of calcineurin signaling, may play a role in myofibrillogenesis.Specifications
| MYOZ1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP98 | |
| MYOZ1 | |
| Human recombit protein fragment corresponding to amino acids 106-299 of human MYOZ1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8A1 | |
| Unconjugated | |
| MYOZ1 | |
| CS-2, FATZ, MYOZ | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 58529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction