missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myozenin 1 Antibody (OTI4C2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72865F
This item is not returnable.
View return policy
Description
Myozenin 1 Monoclonal specifically detects Myozenin 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Myozenin 1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MYOZ1 | |
| Recombinant protein fragment corresponding to amino acids 106-299 of human Myozenin 1 (NP_067068) produced in E.coli. | |
| 0.1 mL | |
| 58529.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4C2 | |
| Immunohistochemistry | |
| calsarcin-2, CS-2, FATZ, Filamin-, actinin- and telethonin-binding protein, MYOZ, myozenin, myozenin 1, myozenin-1, Protein FATZ | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction