missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYPN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570479
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
Striated muscle in vertebrates comprises large proteins which must be organized properly to contract efficiently. Z-lines in striated muscle are a sign of this organization, representing the ends of actin thin filaments, titin, nebulin or nebulette and accessory proteins required for structure and function. This gene encodes a protein which interacts with nebulin in skeletal muscle or nebulette in cardiac muscle and alpha-actinin. In addition, this gene product can interact with a protein with the I-band indicating it has a regulatory as well as structural function. Alternative splicing results in multiple transcript variants.
Specifications
| MYPN | |
| Polyclonal | |
| Unconjugated | |
| MYPN | |
| 1110056A04Rik; 145 kDa sarcomeric protein; 9830112J03; AI853556; CMD1DD; CMH22; Kiaa4170; mKIAA4170; MYOP; Myopalladin; MYPN; RCM4; sarcomeric protein myopalladin, 145 kDa (MYOP); wu:fc17a02 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84665 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86TC9 | |
| MYPN | |
| synthetic peptide directed towards the middle region of human MYPN (aa 972-1021). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction