missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYPOP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571458
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
Transcriptional repressor; DNA-binding protein that specifically recognizes the core sequence 5'-YAAC[GT]G-3'. Dimerization with PFN1 reduces its DNA-binding capacity.
Specifications
| MYPOP | |
| Polyclonal | |
| Unconjugated | |
| MYPOP | |
| Myb related transcription factor, partner of profilin; myb-related protein p42POP; Myb-related transcription factor, partner of profilin; MYPOP; p42 Myb-related transcription factor, partner of profilin; P42pop; Partner of profilin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 339344 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86VE0 | |
| MYPOP | |
| synthetic peptide directed towards the C terminal region of human MYPOP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction