missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYPOP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566761
This item is not returnable.
View return policy
Description
Immunogen sequence: ACPQRYVLSED RREDRRADTS AHSKAGSSSP EPWARPSCTP QEGGCPRPKE RESPP Highest antigen sequence identity to the following orthologs - mouse 71%, rat 73%.
Transcriptional repressor; DNA-binding protein that specifically recognizes the core sequence 5'-YAAC[GT]G-3'. Dimerization with PFN1 reduces its DNA-binding capacity.
Specifications
| MYPOP | |
| Polyclonal | |
| Unconjugated | |
| MYPOP | |
| Myb related transcription factor, partner of profilin; myb-related protein p42POP; Myb-related transcription factor, partner of profilin; MYPOP; p42 Myb-related transcription factor, partner of profilin; P42pop; Partner of profilin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VE0 | |
| MYPOP | |
| Recombinant Human MYPOP. Recombinant protein control fragment (Product #RP-107419). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction