missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYRIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 433.00 - € 588.00
Specifications
| Antigen | MYRIP |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456710
|
Novus Biologicals
NBP1-84137-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18407810
|
Novus Biologicals
NBP1-84137 |
0.1 mL |
€ 588.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYRIP Polyclonal antibody specifically detects MYRIP in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| MYRIP | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol | |
| 25924 | |
| IgG | |
| Immunogen affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SALPSWKSVDRLDETNLAPVLQSPDGNWVALKDGAPPPTRLLAKPKSGTFQALEVASSVASAYDEMGSDSEEDFDWSEALSKLCPRSRALPRNPQPQPTQAQSSDQGPIAASPSSALSPNPEAMCSDSETSSAGSSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title