missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYRIP Polyclonal Antibody

Product Code. 16349214
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16349214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16349214 Supplier Invitrogen™ Supplier No. PA582250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SALPSWKSVD RLDETNLAPV LQSPDGNWVA LKDGAPPPTR LLAKPKSGTF QALEVASSVA SAYDEMGSDS EEDFDWSEAL SKLCPRSRAL PRNPQPQPTQ AQSSDQGPIA ASPSSALSPN PEAMCSDSET SSAGSSR Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 77%.

MYRIP is a protein coding gene. Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor proteins MYO5A and MYO7A. May link RAB27A-containing vesicles to actin filaments. Functions as a protein kinase A-anchoring protein (AKAP). May act as a scaffolding protein that links PKA to components of the exocytosis machinery, thus facilitating exocytosis, including insulin release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYRIP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYRIP
Gene Accession No. Q8NFW9
Gene Alias A230081N12Rik; DKFZp586F1018; exophilin; exophilin 8; exophilin-8; FLJ44025; MGC130034; MGC130035; myosin VIIA and Rab interacting protein; myosin-VIIa- and Rab-interacting protein; MYRIP; OTTHUMP00000209206; Rab effector MyRIP; Slac2c; slaC2-c; slp homolog lacking C2 domains c; Slp homologue lacking C2 domains; synaptotagmin-like protein homologue lacking C2 domains-c; synaptotagmin-like protein lacking C2 domains C
Gene Symbols MYRIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYRIP. Recombinant protein control fragment (Product #RP-96787).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.