missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55252DL594
This item is not returnable.
View return policy
Description
MYST1 Polyclonal antibody specifically detects MYST1 in Human samples. It is validated for Western Blot
Specifications
| MYST1 | |
| Polyclonal | |
| Western Blot | |
| FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1 | |
| The immunogen recognized by this antibody maps to a region between residue 417 and the C-terminus (residue 467) of human males absent on the first (MYST histone acetyltransferase 1) using the numbering given in entry BAB13924.1 (GeneID 84148). | |
| 0.1 mL | |
| Chromatin Research, DNA Repair | |
| 84148 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction