missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYST1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12699-50ul
This item is not returnable.
View return policy
Description
MYST1 Polyclonal antibody specifically detects MYST1 in Human samples. It is validated for Western Blot
Specifications
| MYST1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1 | |
| Produced in rabbits immunized with E. coli-derived Human MYST1 fragment | |
| 50 μg | |
| Chromatin Research, DNA Repair | |
| 84148 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction