Learn More
Invitrogen™ MYST1 Recombinant Rabbit Monoclonal Antibody (6U5Z6)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA538064
Product Code. 17206943
  • € 532.00 / 100µL
Description
Positive Samples: HepG2, MCF7, NIH/3T3 Immunogen sequence: EKPLSDLGKL SYRSYWSWVL LEILRDFRGT LSIKDLSQMT SITQNDIIST LQSLNMVKYW KGQHVICVTP KLVEEHLKSA QYKKPPITVD SVCLKWAPPK HKQVKLSKK.
The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MYST1
Recombinant Monoclonal
1.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D1P2, Q9H7Z6
KAT8
Recombinant fusion protein containing a sequence corresponding to amino acids 350-458 of human KAT8/MYST1/MOF (Q9H7Z6).
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
6U5Z6
Unconjugated
KAT8
2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase Myst1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8
Rabbit
Affinity chromatography
RUO
67773, 84148
-20°C, Avoid Freeze/Thaw Cycles