missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MZF1 Antibody (PCRP-MZF1-1E8) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24013JF635
This item is not returnable.
View return policy
Description
MZF1 Monoclonal antibody specifically detects MZF1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MZF1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| myeloid zinc finger 1, MZF1B, MZF-1zinc finger protein 42 (myeloid-specific retinoic acid-responsive), Zfp98, Zinc finger protein 42, ZNF42MZF, ZSCAN6Zinc finger and SCAN domain-containing protein 6 | |
| Recombinant fragment (around aa37-128) of human MZF1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PCRP-MZF1-1E8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7593 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction