missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,1-Dimethyl-4-(methylamino)-1H-imidazole-5-carboxamide Nitrate (Caffeidine Nitrate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N,1-Dimethyl-4-(methylamino)-1H-imidazole-5-carboxamide Nitrate (Caffeidine Nitrate)
Specifications
Specifications
| Chemical Name or Material | N,1-Dimethyl-4-(methylamino)-1H-imidazole-5-carboxamide Nitrate (Caffeidine Nitrate) |
| CAS | 209725-34-8 |
| Analyte or Component Names | N,1-Dimethyl-4-(methylamino)-1H-imidazole-5-carboxamide Nitrate (Caffeidine Nitrate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H12 N4 O . H N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N,1-Dimethyl-4-(methylamino)-1H-imidazole-5-carboxamide Nitrate, Caffeine Imp. E (EP) as Nitrate, Caffeidine Nitrate, Caffeine Impurity E as Nitrate |
| InChI Formula | InChI=1S/C7H12N4O.HNO3/c1-8-6-5(7(12)9-2)11(3)4-10-6;2-1(3)4/h4,8H,1-3H3,(H,9,12);(H,2,3,4) |
| SMILES | CNC(=O)c1c(NC)ncn1C.O[N+](=O)[O-] |
| IUPAC Name | N,3-dimethyl-5-(methylamino)imidazole-4-carboxamide;nitric acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction