missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, Honeywell Fluka™

Catalog Number p-7145709
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1465-25-4
C12H16Cl2N2
259.17
MZNYWPRCVDMOJG-UHFFFAOYSA-N
15106
CHEBI:53452
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
This item is not returnable. View return policy
1465-25-4
C12H16Cl2N2
259.17
MZNYWPRCVDMOJG-UHFFFAOYSA-N
15106
CHEBI:53452
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

ACS reagent, >98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Packaging Plastic Bottle
Quantity 25 g
Percent Purity min. 98 %
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'