Learn More
N-[(1R)-1-(4-Acetylphenyl)ethyl]acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-[(1R)-1-(4-Acetylphenyl)ethyl]acetamide |
| CAS | 88146-37-6 |
| Molecular Formula | C12H15NO2 |
| Quantity | 100 mg |
| Synonym | (+)-N-[1-(4-Acetylphenyl)ethyl]acetamide |
| InChI Formula | InChI=1S/C12H15NO2/c1-8(13-10(3)15)11-4-6-12(7-5-11)9(2)14/h4-8H,1-3H3,(H,13,15)/t8-/m1/s1 |
| SMILES | C[C@@H](NC(=O)C)c1ccc(cc1)C(=O)C |
| IUPAC Name | N-[(1R)-1-(4-acetylphenyl)ethyl]acetamide |
| Molecular Weight (g/mol) | 205.25 |
| Formula Weight | 205.1103 |