missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride (rac-Demethylselegiline Hydrochloride), Mikromol™
Description
N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride (rac-Demethylselegiline Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | rac-N-Desmethylselegiline Hydrochloride |
| CAS | 2588-96-7 |
| Analyte or Component Names | rac-N-Desmethylselegiline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H15 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride, Selegiline Hydrochloride Imp. D (EP) as Racemate Hydrochloride, rac-Demethylselegiline Hydrochloride, rac-N-Desmethylselegiline Hydrochloride, Selegiline Hydrochloride Imp. D (EP), Selegiline Imp. D (EP), Selegiline Hydrochloride Impurity D as Racemate Hydrochloride, Selegiline Impurity D as Racemate Hydrochloride |
| InChI Formula | InChI=1S/C12H15N.ClH/c1-3-9-13-11(2)10-12-7-5-4-6-8-12;/h1,4-8,11,13H,9-10H2,2H3;1H |
| SMILES | Cl.CC(Cc1ccccc1)NCC#C |
| IUPAC Name | 1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?