Learn More
N,2,4-Trimethylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | N,2,4-Trimethylaniline |
| CAS | 13021-13-1 |
| Molecular Formula | C9 H13 N |
| Quantity | 1 g |
| Synonym | Benzenamine','N,2,4-trimethyl-, 2,4-Xylidine','N-methyl- (6CI,7CI,8CI), N,2,4-Trimethylbenzenamine, 2,4-Dimethyl-N-methylaniline, N,2,4-Trimethylaniline, N-Methyl-2,4-dimethylaniline, N-Methyl-2,4-xylidine |
| InChI Formula | InChI=1S/C9H13N/c1-7-4-5-9(10-3)8(2)6-7/h4-6,10H,1-3H3 |
| SMILES | CNc1ccc(C)cc1C |
| IUPAC Name | N,2,4-trimethylaniline |
| Molecular Weight (g/mol) | 135.21 |
| Formula Weight | 135.1048 |