missing translation for 'onlineSavingsMsg'
Learn More

N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine (Ranitidine Sulphoxide), Mikromol™

Product Code. 17930516
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17930516

Brand: Mikromol MM0086.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine (Ranitidine Sulphoxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ranitidine Sulfoxide
CAS 73851-70-4
Analyte or Component Names Ranitidine Sulfoxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C13 H22 N4 O4 S
Certifications/Compliance ISO 17025
Synonym Ranitidine Hydrochloride Imp. C (EP), Ranitidine Imp. C (EP), Ranitidine USP Related Compound C, Ranitidine USP RC C, N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Ranitidine Sulphoxide, AH 20319X, USP Ranitidine Related Compound C, Ph Eur Ranitidine Impurity C, Ranitidine S-oxide (AH20319), S-oxide (AH 20319X), Ranitidine Related Compound C, Ranitidine Hydrochloride Impurity C, Ranitidine Impurity C
InChI Formula InChI=1S/C13H22N4O4S/c1-14-13(9-17(18)19)15-6-7-22(20)10-12-5-4-11(21-12)8-16(2)3/h4-5,9,14-15H,6-8,10H2,1-3H3
SMILES CNC(=C[N+](=O)[O-])NCCS(=O)Cc1oc(CN(C)C)cc1
IUPAC Name 1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
Molecular Weight (g/mol) 330.4
Quantity 100 mg
Formula Weight 330.1362 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.