Learn More
N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine, TRC
Product Code. 30011518
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011518 25 mg 25mg
30003887 10 mg 10mg
30003749 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011518 Supplier Toronto Research Chemicals Supplier No. TRCE92651025MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1418095-19-8
Molecular Formula C14 H19 N O6 S
Molecular Weight (g/mol) 329.37
Synonym (S)-2-(N-(2-Ethyl-6-methylphenyl)-2-sulfoacetamido)propanoic Acid, N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine
IUPAC Name (2S)-2-(2-ethyl-6-methyl-N-(2-sulfoacetyl)anilino)propanoic acid
SMILES CCc1cccc(C)c1N([C@@H](C)C(=O)O)C(=O)CS(=O)(=O)O
Specifications
Analyte or Component Names N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H19NO6S/c1-4-11-7-5-6-9(2)13(11)15(10(3)14(17)18)12(16)8-22(19,20)21/h5-7,10H,4,8H2,1-3H3,(H,17,18)(H,19,20,21)/t10-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Formula Weight 329.0933
Chemical Name or Material N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine