missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)formamide, 97%

Product Code. 15439007
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15439007

Brand: Thermo Scientific Alfa Aesar H52345.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-06-1 | C3H7NO2 | 89.09 g/mol

N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)formamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-06-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00021040
InChI Key BAMUPQJDKBGDPU-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl formamide, formamide, n-2-hydroxyethyl, n-formylethanolamine
PubChem CID 69657
IUPAC Name N-(2-hydroxyethyl)formamide
SMILES OCCNC=O

Specifications

Linear Formula HCONHCH2CH2OH
Quantity 1 g
Formula Weight 89.09
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxyethyl)formamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.