Learn More
N-2 Trityl Olmesartan Medoxomil, TRC
Specifications
Specifications
| Chemical Name or Material | N-2 Trityl Olmesartan Medoxomil |
| CAS | 1020157-01-0 |
| Molecular Formula | C48 H44 N6 O6 |
| Quantity | 1 g |
| Synonym | 1H-Imidazole-5-carboxylic acid','4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-','(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[(2-triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylate, N-2 Trityl Olmesartan Medoxomil |
| InChI Formula | InChI=1S/C48H44N6O6/c1-5-17-41-49-43(47(3,4)57)42(45(55)58-31-40-32(2)59-46(56)60-40)53(41)30-33-26-28-34(29-27-33)38-24-15-16-25-39(38)44-50-52-54(51-44)48(35-18-9-6-10-19-35,36-20-11-7-12-21-36)37-22-13-8-14-23-37/h6-16,18-29,57H,5,17,30-31H2,1-4H3 |
| SMILES | CCCc1nc(c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn(n5)C(c6ccccc6)(c7ccccc7)c8ccccc8)C(C)(C)O |
| IUPAC Name | (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate |
| Molecular Weight (g/mol) | 800.9 |
| Formula Weight | 800.3322 |