missing translation for 'onlineSavingsMsg'
Learn More

N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-2-naphthalenemethanamine Hydrochloride, TRC

Product Code. 30134760
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134760 500 mg 500mg
17964915 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134760 Supplier Toronto Research Chemicals Supplier No. TRCD473210500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877265-30-0
Molecular Formula C21 H25 N . Cl H
Molecular Weight (g/mol) 327.89
Synonym (2E)-N,6,6-Trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. C (EP) as Hydrochloride, trans-Isoterbinafine Hydrochloride
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMILES Cl.CN(C\C=C\C#CC(C)(C)C)Cc1ccc2ccccc2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H25N.ClH/c1-21(2,3)14-8-5-9-15-22(4)17-18-12-13-19-10-6-7-11-20(19)16-18;/h5-7,9-13,16H,15,17H2,1-4H3;1H/b9-5+;
Formula Weight 327.1754
Percent Purity >95%
Chemical Name or Material trans-Isoterbinafine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.