missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
N-[3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine
Specifications
Specifications
| Chemical Name or Material | N-[3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine |
| CAS | 2724839-84-1 |
| Analyte or Component Names | N-[3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H37 N3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Trimipramine Maleate Imp. E (EP), Trimipramine Imp. E (EP), N-[3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine, Trimipramine Maleate Impurity E, Trimipramine Impurity E |
| InChI Formula | InChI=1S/C21H25NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-13,18,22-23H,2,14-16H2,1H3;1H/b13-12+; |
| SMILES | CC(CN(C)C)CN(C)CC(C)CN1c2ccccc2CCc3ccccc13 |
| IUPAC Name | N-[3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-2-methylpropyl]-N,N',N',2-tetramethylpropane-1,3-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction