missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenylsulfonyl]glycine, 96%, Thermo Scientific™

Product Code. 15438835
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438835 250 mg 250mg
15407157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438835 Supplier Thermo Scientific Alfa Aesar Supplier No. H34214.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85845-02-9
Molecular Formula C9H8F3NO4S
Molecular Weight (g/mol) 283.221
MDL Number MFCD03786832
InChI Key YSZHCOBVLXGMJQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenyl sulfonyl amino acetic acid, 2-3-trifluoromethyl phenylsulfonamido acetic acid, 2-3-trifluoromethyl benzenesulfonamido acetic acid, glycine,n-3-trifluoromethyl phenyl sulfonyl, 3-trifluoromethyl benzenesulfonamido acetic acid, 2-3-trifluoromethyl phenyl sulfonylamino acetic acid, n-3-trifluoromethyl phenylsulfonyl glycine, n-3-trifluoromethyl phenyl sulfonyl glycine, 3-trifluoromethyl benzenesulfonamidoacetic acid
PubChem CID 697902
IUPAC Name 2-[[3-(trifluoromethyl)phenyl]sulfonylamino]acetic acid
SMILES C1=CC(=CC(=C1)S(=O)(=O)NCC(=O)O)C(F)(F)F

Specifications

Melting Point 148°C to 149°C
Quantity 250 mg
Formula Weight 283.23
Percent Purity 96%
Chemical Name or Material N-[3-(Trifluoromethyl)phenylsulfonyl]glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.