Learn More
N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide, Mikromol™
Product Code. 17953936
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 17953936
Brand: Mikromol MM0064.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
CAS 41526-21-0
Analyte or Component Names N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
Shipping Condition Room Temperature
Molecular Formula C14 H10 Br Cl N2 O2
Certifications/Compliance ISO 17025
Synonym N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide, Bromazepam Imp. B (EP), Bromazepam Impurity B
InChI Formula InChI=1S/C14H10BrClN2O2/c15-9-4-5-11(18-13(19)8-16)10(7-9)14(20)12-3-1-2-6-17-12/h1-7H,8H2,(H,18,19)
SMILES ClCC(=O)Nc1ccc(Br)cc1C(=O)c2ccccn2
IUPAC Name N-[4-bromo-2-(pyridine-2-carbonyl)phenyl]-2-chloroacetamide
Molecular Weight (g/mol) 353.6
Quantity 25 mg
Formula Weight 351.9614 g/mol