Learn More
N-(4-Bromophenyl)-[1,1-biphenyl]-4-amine, TRC
Specifications
Specifications
| Chemical Name or Material | N-(4-Bromophenyl)-[1,1-biphenyl]-4-amine |
| CAS | 1160294-93-8 |
| Molecular Formula | C18H14BrN |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C18H14BrN/c19-16-8-12-18(13-9-16)20-17-10-6-15(7-11-17)14-4-2-1-3-5-14/h1-13,20H |
| SMILES | Brc1ccc(Nc2ccc(cc2)c3ccccc3)cc1 |
| IUPAC Name | N-(4-bromophenyl)-4-phenylaniline |
| Molecular Weight (g/mol) | 324.22 |
| Formula Weight | 323.031 |