Learn More
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-oxopropanamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-oxopropanamide |
| CAS | 87310-69-8 |
| Molecular Formula | C11 H7 F3 N2 O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C11H7F3N2O2/c1-6(17)10(18)16-8-3-2-7(5-15)9(4-8)11(12,13)14/h2-4H,1H3,(H,16,18) |
| SMILES | CC(=O)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F |
| IUPAC Name | N-[4-cyano-3-(trifluoromethyl)phenyl]-2-oxopropanamide |
| Molecular Weight (g/mol) | 256.18 |
| Formula Weight | 256.046 |