missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorobenzyl)-3-methylaniline, 97%, Thermo Scientific™

Catalog Number 15422638
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1019518-18-3
C14H14FN
215.271
MFCD11138269
IMKKJCAMRXSOQM-UHFFFAOYSA-N
n-4-fluorobenzyl-3-methylaniline, n-4-fluorophenyl methyl-3-methylaniline
28422119
N-[(4-fluorophenyl)methyl]-3-methylaniline
CC1=CC(=CC=C1)NCC2=CC=C(C=C2)F
This item is not returnable. View return policy
1019518-18-3
C14H14FN
215.271
MFCD11138269
IMKKJCAMRXSOQM-UHFFFAOYSA-N
n-4-fluorobenzyl-3-methylaniline, n-4-fluorophenyl methyl-3-methylaniline
28422119
N-[(4-fluorophenyl)methyl]-3-methylaniline
CC1=CC(=CC=C1)NCC2=CC=C(C=C2)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In materials science, organofluorine compounds have been employed in many niche applications. Fluid fluoropolymers, having a low coefficient of friction, find use as specialty lubricants.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 215.27
Percent Purity 97%
Chemical Name or Material N-(4-Fluorobenzyl)-3-methylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'