missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-n-Butylphenyl)-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65950.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15440037

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD28100496

Specifications

N-(4-n-Butylphenyl)-4-chloro-3-sulfamoylbenzamide
MFCD28100496
Solid
1g
366.86
95%
Videos
Documents