missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Piperidyl)benzamide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-(4-Piperidyl)benzamide
Specifications
Specifications
| Chemical Name or Material | N-(4-Piperidyl)benzamide |
| CAS | 33953-37-6 |
| Analyte or Component Names | N-(4-Piperidyl)benzamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzamide, N-4-piperidinyl-, Benzamide, N-4-piperidyl- (8CI), 4-(Phenylcarboxamido)piperidine, 4-Benzamidopiperidine, N-(4-Piperidinyl)benzamide |
| InChI Formula | InChI=1S/C12H16N2O/c15-12(10-4-2-1-3-5-10)14-11-6-8-13-9-7-11/h1-5,11,13H,6-9H2,(H,14,15) |
| SMILES | O=C(NC1CCNCC1)c2ccccc2 |
| IUPAC Name | N-piperidin-4-ylbenzamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction