Learn More
N-[4-[[(Z)-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)phenylmethyl]amino]phenyl]-N,4-dimethyl-1-piperazineacetamide, TRC
Specifications
Specifications
| CAS | 262366-32-5 |
| Molecular Formula | C29H31N5O2 |
| Quantity | 25 mg |
| Synonym | Desmethoxycarbonyl Nintedanib, Nintedanib Impurity E |
| InChI Formula | InChI=1S/C29H31N5O2/c1-32-16-18-34(19-17-32)20-26(35)33(2)23-14-12-22(13-15-23)30-28(21-8-4-3-5-9-21)27-24-10-6-7-11-25(24)31-29(27)36/h3-15,31,36H,16-20H2,1-2H3 |
| SMILES | CN(C(=O)CN1CCN(C)CC1)c2ccc(N\C(=C\3/C(=O)Nc4ccccc34)\c5ccccc5)cc2 |
| Molecular Weight (g/mol) | 481.59 |
| Chemical Name or Material | N-[4-[((Z)-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)phenylmethyl)amino]phenyl]-N,4-dimethyl-1-piperazineacetamide |