Learn More
N-((4aR,6S,7R,8R,8aS)-6-(Benzyloxy)-8-hydroxy-2-phenylhexahydropyrano[3,2-d][1,3]dioxin-7-yl)acetamide, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 13343-63-0 |
| Molecular Formula | C22H25NO6 |
| MDL Number | MFCD00143679 |
| Quantity | 25 g |
| SMILES | CC(N[C@@H]1[C@@H](O)[C@]2([H])OC(C3=CC=CC=C3)OC[C@@]2([H])O[C@@H]1OCC4=CC=CC=C4)=O |
| Molecular Weight (g/mol) | 399.44 |
| Percent Purity | 95% |
| Chemical Name or Material | N-((4aR,6S,7R,8R,8aS)-6-(Benzyloxy)-8-hydroxy-2-phenylhexahydropyrano[3,2-d][1,3]dioxin-7-yl)acetamide |