missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-Glucosamine Kinase/NAGK Antibody (4D12), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22554AF405
This item is not returnable.
View return policy
Description
N-Acetyl-D-Glucosamine Kinase/NAGK Monoclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| N-Acetyl-D-Glucosamine Kinase/NAGK | |
| Monoclonal | |
| Alexa Fluor 405 | |
| EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910 | |
| Recombinant human N-Acetyl-D-Glucosamine Kinase/NAGK (1-344aa) purified from E. coli | |
| 0.1 mL | |
| metabolism | |
| 55577 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 4D12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction