missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-Glucosamine Kinase/NAGK Antibody (4D12), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-22554R

Product Code. 18412588

  • € 527.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

N-Acetyl-D-Glucosamine Kinase/NAGK Monoclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

N-Acetyl-D-Glucosamine Kinase/NAGK
Monoclonal
DyLight 550
EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910
Recombinant human N-Acetyl-D-Glucosamine Kinase/NAGK (1-344aa) purified from E. coli
0.1 mL
metabolism
55577
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
4D12
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.