missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-arginine, MedChemExpress
Description
N-Acetyl-L-arginine (Ac-Arg-OH) is one of the guanidino compounds found elevated in the serum of an hemodialyzed renal insufficient (uremic) pediatric population.
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-arginine |
| CAS | 155-84-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C8H16N4O3 |
| Synonym | Ac-Arg-OH |
| Solubility Information | H2O : 50 mg/mL (231.22 mM; Need ultrasonic) ∣DMSO : 14.29 mg/mL (66.08 mM; Need ultrasonic) |
| SMILES | O=C(N[C@@H](CCCNC(N)=N)C(O)=O)C |
| Molecular Weight (g/mol) | 216.24 |
| Quantity | 100 mg |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?