missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

Catalog Number 15457649
Click to view available options
Quantity:
1mg
5mg
This item is not returnable. View return policy
292633-16-0
This item is not returnable. View return policy
292633-16-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Content And Storage Store at -20°C
Format Lyophilized Powder
Molecular Weight (g/mol) 711.73
Product Type N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin
Quantity 1mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'