missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-N-phenyl-2,6-dichloroaniline, Mikromol™

Product Code. 30052058
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052058 25 mg 25mg
30049557 100 mg 100mg
30461440 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052058 Supplier Mikromol Supplier No. MM0006.270025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Acetyl-N-phenyl-2,6-dichloroaniline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-N-phenyl-2,6-dichloroaniline
CAS 84803-53-2
Analyte or Component Names N-Acetyl-N-phenyl-2,6-dichloroaniline
Shipping Condition Room Temperature
Molecular Formula C14 H11 Cl2 N O
Certifications/Compliance ISO 17025
Synonym N-Acetyl-N-phenyl-2,6-dichloroaniline
InChI Formula InChI=1S/C14H11Cl2NO/c1-10(18)17(11-6-3-2-4-7-11)14-12(15)8-5-9-13(14)16/h2-9H,1H3
SMILES CC(=O)N(c1ccccc1)c2c(Cl)cccc2Cl
IUPAC Name N-(2,6-dichlorophenyl)-N-phenylacetamide
Molecular Weight (g/mol) 280.15
Quantity 25 mg
Formula Weight 279.0218 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.