missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Sulfadiazine, TRC

Product Code. 30138020
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138020 100 mg 100mg
17978679 10 mg 10mg
17984215 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30138020 Supplier Toronto Research Chemicals Supplier No. TRCA187810100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-74-2
Molecular Formula C12 H12 N4 O3 S
Molecular Weight (g/mol) 292.31
Synonym N-[4-(Pyrimidin-2-ylsulfamoyl)phenyl]acetamide, Sulfadiazine Imp. E (Pharmeuropa), Acetylsulfadiazine
IUPAC Name N-[4-(pyrimidin-2-ylsulfamoyl)phenyl]acetamide
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2ncccn2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H12N4O3S/c1-9(17)15-10-3-5-11(6-4-10)20(18,19)16-12-13-7-2-8-14-12/h2-8H,1H3,(H,15,17)(H,13,14,16)
Formula Weight 292.063
Percent Purity >95%
Chemical Name or Material Acetylsulfadiazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.