missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-trans-4-hydroxy-L-proline, 98%, Thermo Scientific Chemicals

Product Code. p-6909048
Click to view available options
Quantity:
2 g
Unit Size:
2g
This item is not returnable. View return policy

Product Code. 11410218

Brand: Thermo Scientific Alfa Aesar B23767.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33996-33-7
Molecular Formula C7H11NO4
Molecular Weight (g/mol) 173.168
MDL Number MFCD00037339
InChI Key BAPRUDZDYCKSOQ-RITPCOANSA-N
Synonym oxaceprol, n-acetyl-l-hydroxyproline, 2s,4r-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid, oxaceprol inn, ac-hyp-oh, l-proline, 1-acetyl-4-hydroxy-, 4r, unii-q0xv76b96l, ac-hydroxyproline, trans-1-acetyl-4-hydroxy-l-proline, jonctum
PubChem CID 65784
IUPAC Name (2S,4R)-1-acetyl-4-hydroxypyrrolidine-2-carboxylic acid
SMILES CC(=O)N1CC(CC1C(=O)O)O

Specifications

Melting Point 131°C to 133°C
Color White
Beilstein 84043
Merck Index 14,6903
Quantity 2 g
Formula Weight 173.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-trans-4-hydroxy-L-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.