missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic acid, MedChemExpress

Product Code. 16418936
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418936 Supplier Medchem Express Supplier No. HYI0400
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress N-Acetylneuraminic acid is a nine-carbon, sialic acid monosaccharide commonly found in glycoproteins on cell membranes and in glycolipids such as gangliosides in mammalian cells.

N-Acetylneuraminic acid is a nine-carbon, sialic acid monosaccharide commonly found in glycoproteins on cell membranes and in glycolipids such as gangliosides in mammalian cells.

Specifications

Chemical Name or Material N-Acetylneuraminic acid
Physical Form Solid
CAS 131-48-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H19NO9
For Use With (Application) COVID-19-anti-virus
Synonym NANA Lactaminic acid
Solubility Information H2O : ≥ 100 mg/mL (323.34 mM)
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](NC(C)=O)[C@@H](O)CC(C(O)=O)=O
Molecular Weight (g/mol) 309.27
Formula Weight 309.27
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.