missing translation for 'onlineSavingsMsg'
Learn More

N-Allylphthalimide, 97%, Thermo Scientific™

Catalog Number 15400569
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
5428-09-1
MFCD00158662
This item is not returnable. View return policy
5428-09-1
MFCD00158662

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Molecular Formula C11H9NO2
Quantity 250 mg
Synonym 2-allylisoindoline-1,3-dione, n-allylphthalimide, n-2-propenyl phthalimide, 2-allyl-1h-isoindole-1,3 2h-dione, 2-prop-2-en-1-yl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-2-propen-1-yl, 2-prop-2-en-1-yl-2,3-dihydro-1h-isoindole-1,3-dione, n-allylphthalimid, n-allyl-phthalimide, 2-allylisoindole-1,3-dione
InChI Key MHHGQWMCVNQHLG-UHFFFAOYSA-N
SMILES C=CCN1C(=O)C2=CC=CC=C2C1=O
IUPAC Name 2-prop-2-enylisoindole-1,3-dione
Molecular Weight (g/mol) 187.20
PubChem CID 224401
Formula Weight 187.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Allylphthalimide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'