missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine, 99%, Thermo Scientific™

Product Code. 11499487
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499487 1 g 1g
11409497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499487 Supplier Thermo Scientific Alfa Aesar Supplier No. B22010.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMF (0.5 mmol/ml).

Notes
Store at -20°C. Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2389-60-8
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00062271
InChI Key DYSBKEOCHROEGX-GGYSOQFKNA-N
Synonym z-lys boc-oh, n-cbz-n'-boc-l-lysine, nepsilon-boc-nalpha-cbz-l-lysine, cbz-lys boc-oh, z-lysine boc-oh, z-l-lys boc-oh, s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-cbz-n-epsilon-boc-l-lysine, in2-benzyloxycarbonyl-n6-tert-butoxycarbonyl-l-lysine, 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 98765
IUPAC Name (2S)-6-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 64°C to 67°C
Solubility Information Soluble in DMF (0.5 mmol/ml).
Quantity 1 g
Formula Weight 380.44
Percent Purity 99%
Chemical Name or Material N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.