Learn More
N-Benzyl-L-tyrosine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Benzyl-L-tyrosine |
| CAS | 75768-66-0 |
| Molecular Formula | C16H17NO3 |
| Quantity | 50 mg |
| Synonym | N-(Phenylmethyl)-L-tyrosine |
| InChI Formula | InChI=1S/C16H17NO3/c18-14-8-6-12(7-9-14)10-15(16(19)20)17-11-13-4-2-1-3-5-13/h1-9,15,17-18H,10-11H2,(H,19,20)/t15-/m0/s1 |
| SMILES | OC(=O)[C@H](Cc1ccc(O)cc1)NCc2ccccc2 |
| IUPAC Name | (2S)-2-(benzylamino)-3-(4-hydroxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 271.32 |
| Formula Weight | 271.1208 |