Learn More
N-Benzylhydroxylamine hydrochloride, 98%
Product Code. 15453413 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
50g
Product Code. 15453413
missing translation for 'mfr': Thermo Scientific Acros 391040050
Request Bulk or Custom Format
CAS: 29601-98-7 | C7H9NO·ClH | 159.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29601-98-7
Molecular Formula C7H9NO·ClH
Molecular Weight (g/mol) 159.62
InChI Key YSNXOQGDHGUKCZ-UHFFFAOYSA-N
Synonym n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
PubChem CID 11332622
IUPAC Name N-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CNO.Cl
Specifications
Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula C6H5CH2NHOH·HCl
Quantity 5 g
Formula Weight 159.62
Percent Purity 98%
Chemical Name or Material N-Benzylhydroxylamine hydrochloride