Learn More
N-Benzyloxycarbonyl-4-[(3R)-3-amino-1-oxo-4-(phenylthio)butyl]morpholine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Benzyloxycarbonyl-4-[(3R)-3-amino-1-oxo-4-(phenylthio)butyl]morpholine |
| CAS | 870812-93-4 |
| Molecular Formula | C22 H26 N2 O4 S |
| Quantity | 10 mg |
| Synonym | N-[(1R)-3-(4-Morpholinyl)-3-oxo-1-[(phenylthio)methyl]propyl]carbamic Acid Phenylmethyl Ester, [(1R)-3-(4-Morpholinyl)-3-oxo-1-[(phenylthio)methyl]propyl]carbamic Acid Benzyl Ester |
| InChI Formula | InChI=1S/C22H26N2O4S/c25-21(24-11-13-27-14-12-24)15-19(17-29-20-9-5-2-6-10-20)23-22(26)28-16-18-7-3-1-4-8-18/h1-10,19H,11-17H2,(H,23,26)/t19-/m1/s1 |
| SMILES | O=C(N[C@@H](CSc1ccccc1)CC(=O)N2CCOCC2)OCc3ccccc3 |
| IUPAC Name | benzyl N-[(1R)-3-morpholino-3-oxo-1-(phenylsulfanylmethyl)propyl]carbamate |
| Molecular Weight (g/mol) | 414.52 |
| Formula Weight | 414.1613 |