Learn More
N-Benzyloxycarbonyl-L-glutamic acid gamma-tert-butyl ester, 98+%, Thermo Scientific Chemicals
Product Code. 10623025 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10623025 1 g Glass bottle 1g
10366734 5 g Glass bottle 5g
2 options
Product Code. 10623025
Brand: Thermo Scientific Acros 426150010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3886-08-6
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.37
MDL Number MFCD00038274
InChI Key GLMODRZPPBZPPB-ZDUSSCGKSA-M
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
SMILES CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Specifications
CAS Min % 98.0
CAS Max % 100.0
Melting Point 85°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 337.37
Percent Purity 98+%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester