Learn More
N-Biotinyl-N’-maleimido-ethylenediamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Biotinyl-N'-maleimido-ethylenediamine |
| CAS | 139554-72-6 |
| Molecular Formula | C16 H23 N5 O4 S |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C16H23N5O4S/c22-12(17-7-8-18-21-13(23)5-6-14(21)24)4-2-1-3-11-15-10(9-26-11)19-16(25)20-15/h5-6,10-11,15,18H,1-4,7-9H2,(H,17,22)(H2,19,20,25)/t10-,11-,15-/m0/s1 |
| SMILES | O=C(CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NCCNN3C(=O)C=CC3=O |
| IUPAC Name | 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[(2,5-dioxopyrrol-1-yl)amino]ethyl]pentanamide |
| Molecular Weight (g/mol) | 381.45 |
| Formula Weight | 381.1471 |