Learn More
Alfa Aesar™ N-Boc-1-(2,4-dinitrophenyl)-L-histidine, 99%
Merk:  Alfa Aesar™ H63421.22
Overige gegevens : CAS : 25024-53-7 Gewicht : 0.10000kg
Aantal 100g
 Bekijk meerdere types van dit product via de
Produktcode. 15400109
  • € 898.00 / 100 gram
Geschatte verzenddatum
bij leverancier 09-07-2018
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specificatie
Chemische identificatiekenmerken
N-Boc-1-(2,4-dinitrophenyl)-L-histidine
421.36
771922
25024-53-7
MFCD00065966
Boc-His(Dnp)-OH
Specificatie
N-Boc-1-(2,4-dinitrophenyl)-L-histidine
Solid
421.36
771922
100g
25024-53-7
99%
MFCD00065966
Boc-His(Dnp)-OH
Light Sensitive